(3-Acetamidophenyl)boronic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
B23833.03
B23833.06
B23833.14
B23833.03EA
83.7
EUR
B23833.03
B23833.06
B23833.14
(3-Acetamidophenyl)boronic acid ≥98%
(3-Acetamidophenyl)boronic acid
Purity: 98%
CAS No.: 78887-39-5
Molecular Formula: C8H10BNO3
Formula Weight: 178.98
Melting Point: ca 263° dec.
Beilstein Reference: 3278316
Formula:
C₈H₁₀BNO₃ Melting Pt: ∼ 263 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00236013 CAS Number: 78887-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...