β,3-Dinitrostyrene ≥98%

Supplier: Thermo Fisher Scientific
Warning

L12304.03 L12304.06
L12304.03EA 104 EUR
L12304.03 L12304.06
β,3-Dinitrostyrene ≥98%
β,3-Dinitrostyrene

Purity: 98% CAS No.: 882-26-8 Molecular Formula: C8H6N2O4 Formula Weight: 194.15 Melting Point: 125-127° Beilstein Reference: 2051290

Formula: C₈H₆N₂O₄
Melting Pt: 125…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00052063
CAS Number: 882-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR