6-Acetoxy-2-naphthoic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
L15620.06
L15620.06EA
334
EUR
L15620.06
6-Acetoxy-2-naphthoic acid ≥98%
6-Acetoxy-2-naphthoic acid
Purity: 98+%
CAS No.: 17295-26-0
Molecular Formula: C13H10O4
Formula Weight: 230.22
Melting Point: 218-224°
Beilstein Reference: 3310348
Formula:
C₁₃H₁₀O₄ MW: 230,22 g/mol Melting Pt: 218…224 °C Storage Temperature: Ambient |
MDL Number:
MFCD00059556 CAS Number: 17295-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...