α-Bromostyrene 95% stabilised
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
alfa-Bromostyrene, α-Bromostyrene, alpha-Bromostyrene
365800050
365800250
365801000
ACRO365800050EA
90.7
EUR
ACRO365800050
ACRO365800250
ACRO365801000
α-Bromostyrene 95% stabilised
α-Bromostyrene
Formula:
C₈H₇Br MW: 183,0452 g/mol Boiling Pt: 67…70 °C (4 mmHg) Melting Pt: –44 °C Density: 1,41 g/cm³ Flash Pt: 86 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00012229 CAS Number: 98-81-7 EINECS: 202-702-4 |
Specification Test Results
Appearance | Clear colorless to orange Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5870 to 1.5910 (20°C, 589 nm) |
GC | ≥94 % |
Stabilizer | About 0.05% 3,5-di-tert-Butylcatechol |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...