1,2-Ethylenediphosphonic acid ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ethane-1,2-diyldiphosphonic acid

L05828.06 L05828.14
L05828.06EA 107 EUR
L05828.06 L05828.14
1,2-Ethylenediphosphonic acid ≥97%
1,2-Ethylenediphosphonic acid

Purity: 97% CAS No.: 6145-31-9 Molecular Formula: C2H8O6P2 Formula Weight: 190.03 Melting Point: 217-221° Beilstein Reference: 1780250 UN No.: 3261 MDL No.: MFCD00013958

Formula: C₂H₈O₆P₂
MW: 190.03 g/mol
Melting Pt: 217…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00013958
CAS Number: 6145-31-9
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR