2-Chloro-D-phenylalanine ≥98%

Supplier: Thermo Scientific

Synonyms: H-D-Phe(2-Cl)-OH

H27359.03 H27359.MD
H27359.03EA 157 EUR
H27359.03 H27359.MD
2-Chloro-D-phenylalanine ≥98%
2-Chloro-D-phenylalanine

Purity: 98% CAS No.: 80126-50-7 Molecular Formula: C9H10ClNO2 Formula Weight: 199.64 Melting Point: 211-213° MDL No.: MFCD00077920

Formula: C₉H₁₀ClNO₂
MW: 199,64 g/mol
Melting Pt: 211…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00077920
CAS Number: 80126-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR