Benzofuran-2-carboxylic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Benzo[b]furan-2-carboxylic acid

A15579.03 A15579.06 A15579.14
A15579.03EA 40.4 EUR
A15579.03 A15579.06 A15579.14
Benzofuran-2-carboxylic acid ≥98%
Benzofuran-2-carboxylic acid

Purity: 98+% CAS No.: 496-41-3 Molecular Formula: C9H6O3 Formula Weight: 162.15 Melting Point: 192-196° Boiling Point: 310-315° Merck Reference: 14,2561 MDL No.: MFCD00005848

Formula: C₉H₆O₃
MW: 162,15 g/mol
Boiling Pt: 310…315 °C
Melting Pt: 192…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00005848
CAS Number: 496-41-3
EINECS: 207-818-9
Merck Index: 14,2561

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR