Dibenzothiophene-4-boronic acid 95%
Supplier: Thermo Fisher Scientific
|
Warning
|
L19831.03
L19831.06
L19831.03EA
56.8
EUR
L19831.03
L19831.06
Dibenzothiophene-4-boronic acid 95%
Dibenzothiophene-4-boronic acid
Purity: 95%
CAS No.: 108847-20-7
Molecular Formula: C12H9BO2S
Formula Weight: 228.08
Melting Point:
Formula:
C₁₂H₉BO₂S Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318182 CAS Number: 108847-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...