trans-4-Aminocyclohexanecarboxylic acid hydrochloride 96%

Supplier: Thermo Scientific
Warning

H31114.06
H31114.06EA 220 EUR
H31114.06
trans-4-Aminocyclohexanecarboxylic acid hydrochloride 96%
trans-4-Aminocyclohexanecarboxylic acid hydrochloride

Purity: 96% CAS No.: 27960-59-4 Molecular Formula: C7H13NO2.HCl Formula Weight: 179.65 Melting Point: 300° MDL No.: MFCD03844587

Formula: C₇H₁₄ClNO₂
MW: 179,65 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD03844587
CAS Number: 27960-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR