Methyl-1-trifluoromethyl-1-cyclopropanecarboxylate 96%

Supplier: Thermo Scientific
Warning

Synonyms: Methyl 1-(trifluoromethyl)cyclopropanecarboxylate

H25757.03 H25757.06
H25757.03EA 210 EUR
H25757.03 H25757.06
Methyl-1-trifluoromethyl-1-cyclopropanecarboxylate 96%
Methyl 1-trifluoromethyl-1-cyclopropanecarboxylate

Purity: 96% CAS No.: 208242-25-5 Molecular Formula: C6H7F3O2 Formula Weight: 168.12 Flash Point: 42°(108°F) Boiling Point: 115° UN No.: 3272 MDL No.: MFCD06801321

Formula: C₆H₇F₃O₂
MW: 168,12 g/mol
Boiling Pt: 115 °C
Flash Pt: 42 °C (108 °F)
Storage Temperature: Ambient
MDL Number: MFCD06801321
CAS Number: 208242-25-5
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR