1-Cyclohexene-1-carboxylic acid ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Cyclohex-1-enecarboxylic acid
A10741.03
A10741.06
A10741.14
A10741.03EA
43.4
EUR
A10741.03
A10741.06
A10741.14
1-Cyclohexene-1-carboxylic acid ≥97%
1-Cyclohexene-1-carboxylic acid
Purity: 97%
CAS No.: 636-82-8
Molecular Formula: C7H10O2
Formula Weight: 126.16
Flash Point:
Formula:
C₇H₁₀O₂ MW: 126,16 g/mol Boiling Pt: 138…140 °C (14 mmHg) Melting Pt: 30…34 °C Density: 1,100 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001545 CAS Number: 636-82-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...