Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Bis(cycloocta-1,5-dien)rhodium(I)-tetrafluoroborat
375650010
375652500
ACRO375650010EA
827
EUR
ACRO375650010
ACRO375652500
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Bis(1,5-cyclooctadiene)rhodium (I) tetrafluoroborate
Formula:
C₁₆H₂₄BF₄Rh MW: 406,072 g/mol Melting Pt: 210 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00075045 CAS Number: 35138-22-8 EINECS: 460-220-1 UN: 2925 ADR: 4.1,II |
Specification Test Results
Appearance | Red Powder |
Infrared spectrum | Conforms |
Rhodium (Rh) | ≥24.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...