N-(Cyanoacetyl)morpholine ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Morpholino-3-oxopropanenitrile, 3-(Morpholin-4-yl)-3-oxopropanenitrile

A11969.03 A11969.06
A11969.03EA 31.3 EUR
A11969.03 A11969.06
N-(Cyanoacetyl)morpholine ≥98%
N-(Cyanoacetyl)morpholine

Purity: 98% CAS No.: 15029-32-0 Molecular Formula: C7H10N2O2 Formula Weight: 154.17 Odour: Odourless Melting Point: 85-88° Beilstein Reference: 135525 MDL No.: MFCD00023353

Formula: C₇H₁₀N₂O₂
MW: 154,17 g/mol
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00023353
CAS Number: 15029-32-0
EINECS: 239-112-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR