Scopoletin (7-hydroxy-6-methoxycumarin)

Supplier: Roth Carl

Synonyms: Scopoletin, 7-Hydroxy-6-methoxy-2H-chromen-2-one

7210.1 7210.2
ROTH7210.1EA 219 EUR
ROTH7210.1 ROTH7210.2
Scopoletin (7-hydroxy-6-methoxycumarin)
Scopoletin (7-Hydroxy-6-methoxycumarin)
Formula: C₁₀H₈O₄
MW: 192,1681 g/mol
MDL Number: MFCD00006872
CAS Number: 92-61-5
EINECS: 202-171-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR