4-(4-Chlorophenoxy)benzaldehyde ≥98%

Supplier: Thermo Scientific
Warning

H50451.MD
H50451.MDEA 62.5 EUR
H50451.MD
4-(4-Chlorophenoxy)benzaldehyde ≥98%
4-(4-Chlorophenoxy)benzaldehyde

Purity: 98% CAS No.: 61343-99-5 Molecular Formula: C13H9ClO2 Formula Weight: 232.67 MDL No.: MFCD01631895

Formula: C₁₃H₉ClO₂
MW: 232,67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631895
CAS Number: 61343-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR