3,5-Bis(trifluoromethyl)benzyl chloride ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
1-(Chloromethyl)-3,5-bis(trifluoromethyl)benzene
A16553.03
A16553.06
A16553.03EA
41.2
EUR
A16553.03
A16553.06
3,5-Bis(trifluoromethyl)benzyl chloride ≥97%
3,5-Bis(trifluoromethyl)benzyl chloride
Purity: 97%
CAS No.: 75462-59-8
Molecular Formula: C9H5ClF6
Formula Weight: 262.58
Flash Point: 71°(161°F)
Melting Point: 29-33°
Boiling Point: 46°/3mm
Beilstein Reference: 656502
UN No.: 3261
MDL No.: MFCD00009906
Formula:
C₉H₅ClF₆ MW: 262,58 g/mol Boiling Pt: 46 °C (3 mmHg) Melting Pt: 29…33 °C Flash Pt: 71 °C (161 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009906 CAS Number: 75462-59-8 EINECS: 278-216-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...