2-Methoxy-3-nitro-5-pyridineboronic acid pinacol ester 96%

Supplier: Thermo Scientific
Warning

H54933.MD H54933.03
H54933.MDEA 0 EUR
H54933.MD H54933.03
2-Methoxy-3-nitro-5-pyridineboronic acid pinacol ester 96%
2-Methoxy-3-nitro-5-pyridineboronic acid pinacol ester

Purity: 96% CAS No.: 1083168-94-8 Molecular Formula: C12H17BN2O5 Formula Weight: 280.08 Melting Point: 118-119° MDL No.: MFCD12923397

Formula: C₁₂H₁₇BN₂O₅
MW: 280,08 g/mol
Melting Pt: 118…119 °C
Storage Temperature: Ambient
MDL Number: MFCD12923397
CAS Number: 1083168-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR