2-Ethoxy-3-pyridineboronic acid 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(2-Ethoxypyridin-3-yl)boronic acid
L20422.03
L20422.06
L20422.MD
L20422.03EA
159
EUR
L20422.03
L20422.06
L20422.MD
2-Ethoxy-3-pyridineboronic acid 98%
2-Ethoxy-3-pyridineboronic acid
Purity: 98%
CAS No.: 854373-97-0
Molecular Formula: C7H10BNO3
Formula Weight: 166.97
MDL No.: MFCD06201036
Formula:
C₇H₁₀BNO₃ MW: 166,97 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06201036 CAS Number: 854373-97-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...