Methylcyclopentadienylmanganese tricarbonyl ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
47125.03
47125.06
47125.14
47125.22
47125.03EA
45.3
EUR
47125.03
47125.06
47125.14
47125.22
Methylcyclopentadienylmanganese tricarbonyl ≥97%
Methylcyclopentadienylmanganese tricarbonyl
Special handling precautions required. View MSDS prior to purchase.
Insoluble in water. Soluble in hydrocarbon solvents, alcohols, ether, acetone
Purity: 97%
CAS No.: 12108-13-3
Molecular Formula: C9H7MnO3
Formula Weight: 218.09
Flash Point: 96°(205°F)
Physical Form: Liquid
Density: 1.388
Refractive Index: 1.5873
Sensitivity: Air Sensitive
Appearance: Yellow
Melting Point: 1.5°
Boiling Point: 233°
Solubility: Insoluble in water. Soluble in hydrocarbon solvents, alcohols, ether, acetone
Merck Reference: 14,6223
UN No.: 3281
MDL No.: MFCD00001426
Formula:
C₉H₇MnO₃ MW: 218.09 g/mol Boiling Pt: 233 °C Density: 1.388 Flash Pt: 96 °C (205 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001426 CAS Number: 12108-13-3 EINECS: 235-166-5 UN: 3281 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...