5-Amino-2,4-dichloropyrimidine 97%

Supplier: Thermo Scientific
Warning

Synonyms: 2,4-Dichloropyrimidin-5-amine

H32606.06
H32606.06EA 340 EUR
H32606.06
5-Amino-2,4-dichloropyrimidine 97%
5-Amino-2,4-dichloropyrimidine

Purity: 97% CAS No.: 5177-27-5 Molecular Formula: C4H3Cl2N3 Formula Weight: 164.00 MDL No.: MFCD05662684

Formula: C₄H₃Cl₂N₃
MW: 164,00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05662684
CAS Number: 5177-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR