2-Hydroxy-6-methylisonicotinic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Hydroxy-6-methylpyridine-4-carboxylic acid

H27716.03 H27716.06
H27716.03EA 0 EUR
H27716.03 H27716.06
2-Hydroxy-6-methylisonicotinic acid ≥97%
2-Hydroxy-6-methylisonicotinic acid

Purity: 97% CAS No.: 86454-13-9 Molecular Formula: C7H7NO3 Formula Weight: 153.15 Melting Point: 300° dec. MDL No.: MFCD02682019

Formula: C₇H₇NO₃
MW: 153,15 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02682019
CAS Number: 86454-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR