Human Recombinant GPNMB (from HEK293 cells)
Supplier: Antibodies.com
A317722-100 A317722-50
ANTIA317722-100EA 550 EUR
ANTIA317722-100 ANTIA317722-50
Human Recombinant GPNMB (from HEK293 cells)
Partial length Human GPNMB, expressed in HEK293 cells.
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.