Anti-IL-22 Rabbit Monoclonal Antibody [clone: EPR20468-222]

Supplier: Abcam
AB244928-100 AB244928-1
ABCAAB244928-100EA 938 EUR
ABCAAB244928-100 ABCAAB244928-1
Anti-IL-22 Rabbit Monoclonal Antibody [clone: EPR20468-222]
Antibodies
Rabbit monoclonal [EPR20468-222] to IL-22 - BSA and Azide free (Detector).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: IL-22
Clonality: Monoclonal
Clone: EPR20468-222
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR