Anti-Pro-Collagen III alpha 1 Rabbit Recombinant Multiclonal Antibody [clone: RM1028]

Supplier: Abcam
AB283694-100 AB283694-20UL AB283694-1ML
ABCAAB283694-100EA 607 EUR
ABCAAB283694-100 ABCAAB283694-20UL ABCAAB283694-1ML
Anti-Pro-Collagen III alpha 1 Rabbit Recombinant Multiclonal Antibody [clone: RM1028]
Antibodies
Rabbit recombinant multiclonal [RM1028] to Pro-Collagen III alpha 1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Pro-Collagen III alpha 1
Clonality: Recombinant Multiclonal
Clone: RM1028
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR