Anti-PD-L1 Rabbit Monoclonal Antibody [clone: EPR20529]

Supplier: Abcam
AB213480-100 AB213480-1ML
ABCAAB213480-100EA 688 EUR
ABCAAB213480-100 ABCAAB213480-1ML
Anti-PD-L1 Rabbit Monoclonal Antibody [clone: EPR20529]
Antibodies
Rabbit monoclonal [EPR20529] to PD-L1.

Type: Primary
Antigen: PD-L1
Clonality: Monoclonal
Clone: EPR20529
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR