Anti-p57 Kip2 Rabbit Polyclonal Antibody

Supplier: Abcam
AB99888-500 AB99888-1 AB99888-100
ABCAAB99888-500EA 919 EUR
ABCAAB99888-500 ABCAAB99888-1 ABCAAB99888-100
Anti-p57 Kip2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to p57 Kip2.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: p57 Kip2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR