Anti-FGF2 Rabbit Monoclonal Antibody [clone: EPR20145-213]

Supplier: Abcam
AB259599-1 AB259599-100
ABCAAB259599-1EA 9320 EUR
ABCAAB259599-1 ABCAAB259599-100
Anti-FGF2 Rabbit Monoclonal Antibody [clone: EPR20145-213]
Antibodies
Rabbit monoclonal [EPR20145-213] to FGF2 - BSA and Azide free (Capture).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: FGF2
Clonality: Monoclonal
Clone: EPR20145-213
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR