Anti-C5 / C5b Rabbit Monoclonal Antibody [clone: EPR23940-3]

Supplier: Abcam
AB275931-100 AB275931-1ML
ABCAAB275931-100EA 707 EUR
ABCAAB275931-100 ABCAAB275931-1ML
Anti-C5 / C5b Rabbit Monoclonal Antibody [clone: EPR23940-3]
Antibodies
Rabbit monoclonal [EPR23940-3] to C5 / C5b.

Type: Primary
Antigen: C5 / C5b
Clonality: Monoclonal
Clone: EPR23940-3
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR