Anti-TrkA+TrkB+TrkC (phospho Y496) Rabbit Monoclonal Antibody [clone: EPR19140]

Supplier: Abcam
AB251238-1 AB251238-100
ABCAAB251238-1EA 9160 EUR
ABCAAB251238-1 ABCAAB251238-100
Anti-TrkA+TrkB+TrkC (phospho Y496) Rabbit Monoclonal Antibody [clone: EPR19140]
Antibodies
Rabbit monoclonal [EPR19140] to TrkA+TrkB+TrkC (phospho Y496 + Y516 + Y516) - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: TrkA+TrkB+TrkC
Clonality: Monoclonal
Clone: EPR19140
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR