Anti-Dnmt1 Rat Monoclonal Antibody [clone: 2C1]

Supplier: Abcam
AB255781-100 AB255781-1
ABCAAB255781-100EA 1020 EUR
ABCAAB255781-100 ABCAAB255781-1
Anti-Dnmt1 Rat Monoclonal Antibody [clone: 2C1]
Antibodies
Rat monoclonal [2C1] to Dnmt1 - BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Dnmt1
Clonality: Monoclonal
Clone: 2C1
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR