Anti-Carbonic Anhydrase 6/CA6 Rabbit Monoclonal Antibody [clone: EPR13954(2)]

Supplier: Abcam
AB210406-100 AB210406-1ML
ABCAAB210406-100EA 625 EUR
ABCAAB210406-100 ABCAAB210406-1ML
Anti-Carbonic Anhydrase 6/CA6 Rabbit Monoclonal Antibody [clone: EPR13954(2)]
Antibodies
Rabbit monoclonal [EPR13954(2)] to Carbonic Anhydrase 6/CA6.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Carbonic Anhydrase 6/CA6
Clonality: Monoclonal
Clone: EPR13954(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR