Anti-Collagen IV Rabbit Monoclonal Antibody [clone: EPR22911-127]

Supplier: Abcam
AB236640-100 AB236640-1ML
ABCAAB236640-100EA 725 EUR
ABCAAB236640-100 ABCAAB236640-1ML
Anti-Collagen IV Rabbit Monoclonal Antibody [clone: EPR22911-127]
Antibodies
Rabbit monoclonal [EPR22911-127] to Collagen IV.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Collagen IV
Clonality: Monoclonal
Clone: EPR22911-127
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR