Anti-TRMT5 Rabbit Monoclonal Antibody [clone: EPR24139-11]

Supplier: Abcam
AB259986-100 AB259986-20UL AB259986-1ML
ABCAAB259986-100EA 713 EUR
ABCAAB259986-100 ABCAAB259986-20UL ABCAAB259986-1ML
Anti-TRMT5 Rabbit Monoclonal Antibody [clone: EPR24139-11]
Antibodies
Rabbit monoclonal [EPR24139-11] to TRMT5.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TRMT5
Clonality: Monoclonal
Clone: EPR24139-11
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR