Anti-NF-kB p65 (phospho S536) Rabbit Monoclonal Antibody [clone: NFKBp65S536-B7] (FITC (Fluorescein Isothiocyanate))

Supplier: Abcam
AB278631-100 AB278631-10
ABCAAB278631-100EA 819 EUR
ABCAAB278631-100 ABCAAB278631-10
Anti-NF-kB p65 (phospho S536) Rabbit Monoclonal Antibody [clone: NFKBp65S536-B7] (FITC (Fluorescein Isothiocyanate))
Antibodies
FITC Rabbit monoclonal [NFKBp65S536-B7] to NF-kB p65 (phospho S536).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: NF-kB p65
Clonality: Monoclonal
Clone: NFKBp65S536-B7
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR