Anti-TFEB (phospho S211) Rabbit Monoclonal Antibody [clone: EPR27960-64]

Supplier: Abcam
AB310331-100UL AB310331-20UL AB310331-1ML
ABCAAB310331-100ULEA 694 EUR
ABCAAB310331-100UL ABCAAB310331-20UL ABCAAB310331-1ML
Anti-TFEB (phospho S211) Rabbit Monoclonal Antibody [clone: EPR27960-64]
Antibodies
Rabbit monoclonal [EPR27960-64] to TFEB (phospho S211).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: TFEB
Clonality: Monoclonal
Clone: EPR27960-64
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR