Anti-xCT Rabbit Monoclonal Antibody [clone: EPR27115-64]

Supplier: Abcam
AB307601-10 AB307601-100 AB307601-20UL AB307601-1ML
ABCAAB307601-10EA 232 EUR
ABCAAB307601-10 ABCAAB307601-100 ABCAAB307601-20UL ABCAAB307601-1ML
Anti-xCT Rabbit Monoclonal Antibody [clone: EPR27115-64]
Antibodies
Rabbit monoclonal [EPR27115-64] to xCT.

Type: Primary
Antigen: xCT
Clonality: Monoclonal
Clone: EPR27115-64
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR