Anti-Helicobacter pylori Mouse Monoclonal Antibody [clone: SPM526]

Supplier: Abcam
AB231433-100 AB231433-1
ABCAAB231433-100EA 594 EUR
ABCAAB231433-100 ABCAAB231433-1
Anti-Helicobacter pylori Mouse Monoclonal Antibody [clone: SPM526]
Antibodies
Mouse monoclonal [SPM526] to Helicobacter pylori.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Helicobacter pylori
Clonality: Monoclonal
Clone: SPM526
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Helpy
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR