Anti-alpha smooth muscle Actin+ACTG2 (acetyl E3) Rabbit Monoclonal Antibody [clone: E184]

Supplier: Abcam
AB215368-100 AB215368-1
ABCAAB215368-100EA 1020 EUR
ABCAAB215368-100 ABCAAB215368-1
Anti-alpha smooth muscle Actin+ACTG2 (acetyl E3) Rabbit Monoclonal Antibody [clone: E184]
Antibodies
Rabbit monoclonal [E184] to alpha smooth muscle Actin (acetyl E3) + ACTG2 (acetyl E3) - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: alpha smooth muscle Actin+ACTG2
Clonality: Monoclonal
Clone: E184
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR