Anti-NUR77 Rabbit Monoclonal Antibody [clone: EPR23982-85]

Supplier: Abcam
AB283264-100 AB283264-20UL AB283264-1ML
ABCAAB283264-100EA 713 EUR
ABCAAB283264-100 ABCAAB283264-20UL ABCAAB283264-1ML
Anti-NUR77 Rabbit Monoclonal Antibody [clone: EPR23982-85]
Antibodies
Rabbit monoclonal [EPR23982-85] to NUR77.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: NUR77
Clonality: Monoclonal
Clone: EPR23982-85
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR