Anti-GABA A Receptor beta 2/GABRB2 Rabbit Monoclonal Antibody [clone: EPR10517]

Supplier: Abcam
AB156000-100 AB156000-1ML
ABCAAB156000-100EA 619 EUR
ABCAAB156000-100 ABCAAB156000-1ML
Anti-GABA A Receptor beta 2/GABRB2 Rabbit Monoclonal Antibody [clone: EPR10517]
Antibodies
Rabbit monoclonal [EPR10517] to GABA A Receptor beta 2/GABRB2.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: GABA A Receptor beta 2/GABRB2
Clonality: Monoclonal
Clone: EPR10517
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR