Anti-Histone H4 (acetyl K5) Rabbit Monoclonal Antibody [clone: EP1000Y] (Alexa Fluor® 488)

Supplier: Abcam
AB313346-10UL AB313346-100UL
ABCAAB313346-10ULEA 238 EUR
ABCAAB313346-10UL ABCAAB313346-100UL
Anti-Histone H4 (acetyl K5) Rabbit Monoclonal Antibody [clone: EP1000Y] (Alexa Fluor® 488)
Antibodies
Alexa Fluor® 488 Rabbit monoclonal [EP1000Y] to Histone H4 (acetyl K5).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Histone H4
Clonality: Monoclonal
Clone: EP1000Y
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR