Anti-Cathepsin D Chicken chimera Monoclonal Antibody [clone: EPR3057Y-CkIgY]

Supplier: Abcam
AB302647-1 AB302647-100
ABCAAB302647-1EA 9160 EUR
ABCAAB302647-1 ABCAAB302647-100
Anti-Cathepsin D Chicken chimera Monoclonal Antibody [clone: EPR3057Y-CkIgY]
Antibodies
Chicken monoclonal [EPR3057Y-CkIgY] to Cathepsin D - BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cathepsin D
Clonality: Monoclonal
Clone: EPR3057Y-CkIgY
Conjugation:
Epitope:
Host: Chicken chimera
Isotype: IgY
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR