Anti-Sodium Potassium ATPase Rabbit Monoclonal Antibody [clone: EP1845Y] (APC (Allophycocyanin))

Supplier: Abcam
AB314288-100UL AB314288-10UL
ABCAAB314288-100ULEA 819 EUR
ABCAAB314288-100UL ABCAAB314288-10UL
Anti-Sodium Potassium ATPase Rabbit Monoclonal Antibody [clone: EP1845Y] (APC (Allophycocyanin))
Antibodies
APC Rabbit monoclonal [EP1845Y] to Sodium Potassium ATPase - Plasma Membrane Loading Control.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Control
Antigen: Sodium Potassium ATPase
Clonality: Monoclonal
Clone: EP1845Y
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR