Anti-Fast Myosin Skeletal Heavy chain+MYH4 Rabbit Monoclonal Antibody [clone: EPR22880-64]

Supplier: Abcam
AB221149-100 AB221149-20UL AB221149-1ML
ABCAAB221149-100EA 475 EUR
ABCAAB221149-100 ABCAAB221149-20UL ABCAAB221149-1ML
Anti-Fast Myosin Skeletal Heavy chain+MYH4 Rabbit Monoclonal Antibody [clone: EPR22880-64]
Antibodies
Rabbit monoclonal [EPR22880-64] to Fast Myosin Skeletal Heavy chain + MYH4.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Fast Myosin Skeletal Heavy chain+MYH4
Clonality: Monoclonal
Clone: EPR22880-64
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR