Anti-Factor IX/PTC Rabbit Monoclonal Antibody [clone: EPR22846-238]

Supplier: Abcam
AB255824-100 AB255824-1ML
ABCAAB255824-100EA 757 EUR
ABCAAB255824-100 ABCAAB255824-1ML
Anti-Factor IX/PTC Rabbit Monoclonal Antibody [clone: EPR22846-238]
Antibodies
Rabbit monoclonal [EPR22846-238] to Factor IX/PTC.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Factor IX/PTC
Clonality: Monoclonal
Clone: EPR22846-238
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR