Anti-IL-4 Rabbit Monoclonal Antibody [clone: EPR22879-236] (Alexa Fluor® 647)

Supplier: Abcam
AB303475-100 AB303475-10
ABCAAB303475-100EA 775 EUR
ABCAAB303475-100 ABCAAB303475-10
Anti-IL-4 Rabbit Monoclonal Antibody [clone: EPR22879-236] (Alexa Fluor® 647)
Antibodies
Alexa Fluor® 647 Rabbit monoclonal [EPR22879-236] to IL-4.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: IL-4
Clonality: Monoclonal
Clone: EPR22879-236
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR