Anti-Mad2L1 Rabbit Monoclonal Antibody [clone: EPR9852]

Supplier: Abcam
AB171084-100 AB171084-1ML
ABCAAB171084-100EA 663 EUR
ABCAAB171084-100 ABCAAB171084-1ML
Anti-Mad2L1 Rabbit Monoclonal Antibody [clone: EPR9852]
Antibodies
Rabbit monoclonal [EPR9852] to Mad2L1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Mad2L1
Clonality: Monoclonal
Clone: EPR9852
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR