Anti-IL-10RB/IL-10R2 Rabbit Monoclonal Antibody [clone: EPR21829-213]

Supplier: Abcam
AB259505-1 AB259505-100
ABCAAB259505-1EA 9320 EUR
ABCAAB259505-1 ABCAAB259505-100
Anti-IL-10RB/IL-10R2 Rabbit Monoclonal Antibody [clone: EPR21829-213]
Antibodies
Rabbit monoclonal [EPR21829-213] to IL-10RB/IL-10R2 - BSA and Azide free (Capture).

Type: Primary
Antigen: IL-10RB/IL-10R2
Clonality: Monoclonal
Clone: EPR21829-213
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR