Anti-Integrin beta 1 Rabbit Monoclonal Antibody [clone: EP1041Y]

Supplier: Abcam
AB192456-100 AB192456-1
ABCAAB192456-100EA 1020 EUR
ABCAAB192456-100 ABCAAB192456-1
Anti-Integrin beta 1 Rabbit Monoclonal Antibody [clone: EP1041Y]
Antibodies
Rabbit monoclonal [EP1041Y] to Integrin beta 1 - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Integrin beta 1
Clonality: Monoclonal
Clone: EP1041Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR