Anti-Histone H3 (mono methyl K4) Rabbit Monoclonal Antibody [clone: ERP16597]

Supplier: Abcam
AB176877-100 AB176877-1ML
ABCAAB176877-100EA 725 EUR
ABCAAB176877-100 ABCAAB176877-1ML
Anti-Histone H3 (mono methyl K4) Rabbit Monoclonal Antibody [clone: ERP16597]
Antibodies
Rabbit monoclonal [ERP16597] to Histone H3 (mono methyl K4) - ChIP Grade.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: ERP16597
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR